ISAspher HPLC Columns, Amino
Leverancier: ISERA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A111-323A00 A111-323B00 A111-323C00 A111-323D00 A111-323E00 A111-323F00 A111-323G00 A111-323H00 A111-323J00 A111-323K00 A111-323L00 A111-323M00 A111-323N00 A111-323P00 A111-323Q00 A111-323R00 A111-323S00 A111-323T00 A111-323U00 A111-323V00 A111-323W00 A111-323X00 A111-323Y00 A111-323Z00 A111-325A00 A111-325B00 A111-325C00 A111-325D00 A111-325E00 A111-325F00 A111-325G00 A111-325H00 A111-325J00 A111-325K00 A111-325L00 A111-325M00 A111-325N00 A111-325P00 A111-325Q00 A111-325R00 A111-325S00 A111-325T00 A111-325U00 A111-325V00 A111-325W00 A111-325X00 A111-325Y00 A111-325Z00 A111-329A00 A111-329B00 A111-329C00 A111-329D00 A111-329E00 A111-329F00 A111-329G00 A111-329H00 A111-329J00 A111-329K00 A111-329L00 A111-329M00 A111-329N00 A111-329P00 A111-329Q00 A111-329R00 A111-329S00 A111-329T00 A111-329U00 A111-329V00 A111-329W00 A111-329X00 A111-329Y00 A111-329Z00
ISERA111-323A00EA 289 EUR
ISERA111-323A00 ISERA111-323B00 ISERA111-323C00 ISERA111-323D00 ISERA111-323E00 ISERA111-323F00 ISERA111-323G00 ISERA111-323H00 ISERA111-323J00 ISERA111-323K00 ISERA111-323L00 ISERA111-323M00 ISERA111-323N00 ISERA111-323P00 ISERA111-323Q00 ISERA111-323R00 ISERA111-323S00 ISERA111-323T00 ISERA111-323U00 ISERA111-323V00 ISERA111-323W00 ISERA111-323X00 ISERA111-323Y00 ISERA111-323Z00 ISERA111-325A00 ISERA111-325B00 ISERA111-325C00 ISERA111-325D00 ISERA111-325E00 ISERA111-325F00 ISERA111-325G00 ISERA111-325H00 ISERA111-325J00 ISERA111-325K00 ISERA111-325L00 ISERA111-325M00 ISERA111-325N00 ISERA111-325P00 ISERA111-325Q00 ISERA111-325R00 ISERA111-325S00 ISERA111-325T00 ISERA111-325U00 ISERA111-325V00 ISERA111-325W00 ISERA111-325X00 ISERA111-325Y00 ISERA111-325Z00 ISERA111-329A00 ISERA111-329B00 ISERA111-329C00 ISERA111-329D00 ISERA111-329E00 ISERA111-329F00 ISERA111-329G00 ISERA111-329H00 ISERA111-329J00 ISERA111-329K00 ISERA111-329L00 ISERA111-329M00 ISERA111-329N00 ISERA111-329P00 ISERA111-329Q00 ISERA111-329R00 ISERA111-329S00 ISERA111-329T00 ISERA111-329U00 ISERA111-329V00 ISERA111-329W00 ISERA111-329X00 ISERA111-329Y00 ISERA111-329Z00
ISAspher HPLC Columns, Amino
Kolommen voor chromatografie
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µMore Product Information