XELA UHPLC Columns, C18-AQ-PolE
Leverancier: ISERA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A211-Q11A00 A211-Q11B00 A211-Q11D00 A211-Q11F00 A211-Q11G00 A211-Q11H00 A211-Q11K00 A211-Q11M00 A211-Q11U00 A211-Q11V00 A211-Q11X00 A211-Q11Z00 A212-Q11A30
ISERA211-Q11A00EA 476 EUR
ISERA211-Q11A00 ISERA211-Q11B00 ISERA211-Q11D00 ISERA211-Q11F00 ISERA211-Q11G00 ISERA211-Q11H00 ISERA211-Q11K00 ISERA211-Q11M00 ISERA211-Q11U00 ISERA211-Q11V00 ISERA211-Q11X00 ISERA211-Q11Z00 ISERA212-Q11A30
XELA UHPLC Columns, C18-AQ-PolE
Kolommen voor chromatografie
A maximum of separation efficiency and perfect selectivity comply with highest demands.
  • Optimal resolution is achieved by high separation efficiency and appropriate selectivity
  • Powerful
  • Fast and sensitive
  • UHPLC without limitation
XELA columns for UHPLC combine these two crucial parameters of efficency and selectivity in an excellent manner. Therefore, they are the perfect tool for successful LC separations in the high-performance section.
1,7 µm silica particles with a narrow particle size distribution and a homogeneous pore structure ensureMore Product Information