Isofield Blazer LT Sterile Latex Cleanroom Gloves
Leverancier: ISOFIELD IRELAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1063010LT 1063060LT 1063065LT 1063070LT 1063075LT 1063080LT 1063085LT 1063090LT
ISOF1063010LTEA 374 EUR
ISOF1063010LT ISOF1063060LT ISOF1063065LT ISOF1063070LT ISOF1063075LT ISOF1063080LT ISOF1063085LT ISOF1063090LT
Isofield Blazer LT Sterile Latex Cleanroom Gloves
Handschoenen
Isofield Blazer LT sterile latex cleanroom gloves are suitable for use in GMP Grade A and B, ISO Class 4 cleanrooms and above.
  • Hand-specific
  • Natural colour
  • Textured palm and fingers
  • Beaded cuff
  • Low endotoxin levels
  • Protein level less than 50 μg/g
  • Gamma irradiated, minimum 25 kGy
  • Sterility Assurance Level 10⁻⁶
  • IPA resistant ink pouches
This ultra-clean latex glove is triple washed in deionized water and its surface is smooth enough for double-donning whilst also offering a textured palm and fingers for added grip. Every Blazer LT lot is tested for
More Product Information