PCR plates, 96-well, fully skirted, exo-frame
Leverancier: iST Scientific Ltd
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
IST-601-096GCT
ISTSIST-601-096GCTEA 419 EUR
ISTSIST-601-096GCT
PCR plates, 96-well, fully skirted, exo-frame
Microplaten PCR-platen
Fully skirted 96-well PCR plate with clear PP wells and grey PC frame.
  • Free of DNA, DNase, RNase, and PCR inhibitors
  • Low binding PP wells
  • Alpha numberic labeling
  • Manufactured under the most modern clean room conditions
  • Automation friendly
Rigid two-component design eliminates warping and distortion during PCR.
Verpakking: Packaged 10 plates per sleeve, 50 plates per box.