Potassium oxonate

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J60400.06 J60400.14 J60400.22
J60400.06EA 37.9 EUR
J60400.06 J60400.14 J60400.22
Potassium oxonate
Potassium oxonate

CAS No.: 2207-75-2 Molecular Formula: C4H2KN3O4 Formula Weight: 195.18 Physical Form: Powder Appearance: Off-white Melting Point: 300° MDL No.: MFCD00010565


Applications: Inhibitor of uricase

Formule: C₄H₃N₃O₄
Smeltpunt: 300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010565
CAS nummer: 2207-75-2
EINECS: 218-627-5
Merck Index: 14,6947

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR