Tricine-buffered saline (5X), pH 7,0

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J60811.AK
J60811.AKEA 60.8 EUR
J60811.AK
Tricine-buffered saline (5X), pH 7,0
Buffers Biologische buffers

CAS No.: 5704-04-1
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00004277

Order Now

Specificaties

Environmentally Preferable
Geur/lucht Odourless
Physical form Liquid
CAS No. 5704-04-1
MDL No. MFCD00004277
Uiterlijk Colourless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR