TRIS-buffered saline (TBS, 20X) pH 7,4

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J60877.K2 J60877.K3
J60877.K2EA 75 EUR
J60877.K2 J60877.K3
TRIS-buffered saline (TBS, 20X) pH 7,4
Buffers Buffers voor elektroforese Wasbuffers

CAS No.: 77-86-1
Physical Form: Liquid
MDL No.: MFCD00132476

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR