Phenethyl caffeate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

J61386.MD J61386.03
J61386.MDEA 127 EUR
J61386.MD J61386.03
Phenethyl caffeate ≥99%
Phenethyl caffeate

Purity: 99+% CAS No.: 104594-70-9 Molecular Formula: C17H16O4 Formula Weight: 284.31 Storage Temperature: -30°C to -10°C Physical Form: Crystalline powder Sensitivity: Light Sensitive Appearance: White to yellow Melting Point: 127-129° Solubility: Soluble in ethyl acetate at 50mg/ml. Soluble in DMSO and ethanol. MDL No.: MFCD00866470


Applications: Inhibitor of protein tyrosine kinase, and is cytotoxic to cancer cell lines

Formule: C₁₇H₁₆O₄
Smeltpunt: 127…129 °C
MDL Nummer: MFCD00866470
CAS nummer: 104594-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR