Buffers, BICINE, 0,5 M solutioins

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J61632.AE J61632.AK J60494.AE J60494.AK J63924.AE J63924.AK
J61632.AEEA 74.5 EUR
J61632.AE J61632.AK J60494.AE J60494.AK J63924.AE J63924.AK
Buffers, BICINE, 0,5 M solutioins
Buffers Biologische buffers

N,N-(2-Dihydroxyethyl)glycine (BICINE) buffer solutions.


CAS No.: 150-25-4
MDL No.: MFCD00004295


BICINE is used for the preparation of stable substrate solution for serum guanase determination. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.


Opgelet: Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Order Now

Specificaties

Environmentally Preferable
Physical form Liquid Liquid
CAS No. 150-25-4 150-25-4
MDL No. MFCD00004295 MFCD00004295
Opslagtemperatuur 2°C to 8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR