Guanosine-5'-diphosphate disodium salt

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J61646.MCEA 119 EUR
J61646.MC J61646.ME
Guanosine-5'-diphosphate disodium salt
Guanosine-5'-diphosphate disodium salt

CAS No.: 7415-69-2 Molecular Formula: C10H13N5Na2O11P2 Formula Weight: 487.17 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00084665

Formule: C₁₀H₁₃N₅Na₂O₁₁P₂
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00070121
CAS nummer: 7415-69-2
EINECS: 231-026-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR