Sodium alginate, high viscosity

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Alginic acid sodium salt, Algin

J61887.22 J61887.30 J61887.A1
J61887.22EA 65.2 EUR
J61887.22 J61887.30 J61887.A1
Sodium alginate, high viscosity
Sodium alginate

CAS No.: 9005-38-3 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white Merck Reference: 14,241 MDL No.: MFCD00081310


A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studies

Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081310
CAS nummer: 9005-38-3
Merck Index: 14,241

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR