Sodium Cacodylate, 0,1 M Buffer Solution, pH 6,8

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J62202.AEEA 123 EUR
J62202.AE
Sodium Cacodylate, 0,1 M Buffer Solution, pH 6,8
Buffers Biologische buffers

Liquid.


CAS No.: 6131-99-3
Storage Temperature: 2 to 8 ℃
Physical Form: Liquid
MDL No.: MFCD00149079

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR