Buffers, AMPSO, 0,2 M solutions

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J62328.AE J62328.AK
J62328.AEEA 55.5 EUR
J62328.AE J62328.AK
Buffers, AMPSO, 0,2 M solutions
Buffers Biologische buffers

CAS No.: 102029-60-7
Storage temperature: 2 to 8 °C
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00067500

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR