G418 disulphate cell culture reagent

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate

J62671.03 J62671.06 J62671.14
J62671.03EA 113 EUR
J62671.03 J62671.06 J62671.14
G418 disulphate cell culture reagent
G418 disulphate

CAS No.: 108321-42-2 Molecular Formula: C20H40N4O10.2H2SO4 Formula Weight: 692.71 Physical Form: Powder Appearance: White Solubility: Soluble in water at 50mg/ml MDL No.: MFCD00058314


Aminoglycoside that is toxic to bacteria, yeast, protozoans and helminths

Formule: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00058314
CAS nummer: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR