Phosphate-buffered saline (PBS, 10X), pH 7,6

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J62692.K2 J62692.K3 J62692.K7
J62692.K2EA 72.1 EUR
J62692.K2 J62692.K3 J62692.K7
Phosphate-buffered saline (PBS, 10X), pH 7,6
Buffers Buffers voor weefselkweek

Physical Form: Liquid
MDL No.: MFCD00131855

Order Now

Specificaties

Environmentally Preferable
Physical form Liquid
MDL No. MFCD00131855

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR