1-Naphthyl phosphate monosodium salt monohydrate ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-Naphthylphosphoric Acid Monosodium Salt Monohydrate, Monosodium 1-Naphthyl Phosphate Monohydrate, Sodium 1-Naphthyl Hydrogen Phosphate Monohydrate, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate

J63004.06 J63004.14
J63004.06EA 79.6 EUR
J63004.06 J63004.14
1-Naphthyl phosphate monosodium salt monohydrate ≥97%
1-Naphthyl phosphate monosodium salt monohydrate

Purity: 97+% CAS No.: 81012-89-7 Molecular Formula: C10H8NaO4P.H2O Formula Weight: 264.15 (246.13anhy) Storage Temperature: 2°C to 8°C Physical Form: Powder Solubility: Soluble in water Beilstein Reference: 1877764 MDL No.: MFCD00150615


Substrate for determination of phosphatases

Formule: C₁₀H₈NaO₄P·1H₂O
MW: 264.15 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00150615
CAS nummer: 81012-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR