N-(1-Naphthyl)ethylenediamine dihydrochloride ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine

J63214.06 J63214.14 J63214.22
J63214.06EA 43 EUR
J63214.06 J63214.14 J63214.22
N-(1-Naphthyl)ethylenediamine dihydrochloride ACS
N-(1-Naphthyl)ethylenediamine dihydrochloride

CAS No.: 1465-25-4 Molecular Formula: C12H14N2.2HCl Formula Weight: 259.18 Physical Form: Powder Sensitivity: Light Sensitive and Hygroscopic Appearance: White to cream or pale gr Melting Point: 191-199° Solubility: Soluble in water or 95% ethanol Beilstein Reference: 3707471 Merck Reference: 14,6409 MDL No.: MFCD00012556


Used for Colourimetric determination of sulfonamide in blood

Formule: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Smeltpunt: 191…199 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012556
CAS nummer: 1465-25-4
EINECS: 215-981-2
Merck Index: 14,6409
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR