1,1-Dimethylbiguanide hydrochloride

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

J63361.06 J63361.14 J63361.22
J63361.06EA 41.6 EUR
J63361.06 J63361.14 J63361.22
1,1-Dimethylbiguanide hydrochloride
1,1-Dimethylbiguanide hydrochloride

CAS No.: 1115-70-4 Molecular Formula: C4H11N5.HCl Formula Weight: 165.62 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Melting Point: 218-226° Merck Reference: 14,5938 MDL No.: MFCD00012582


An antidiabetic agent

Formule: C₄H₁₂ClN₅
Smeltpunt: 218…226 °C
MDL Nummer: MFCD00012582
CAS nummer: 1115-70-4
EINECS: 214-230-6
Merck Index: 14,5938

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR