Laemmli SDS sample buffer, non-reducing (4X)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J63615.ACEA 61.9 EUR
J63615.AC J63615.AD
Laemmli SDS sample buffer, non-reducing (4X)
Buffers Buffers voor elektroforese Loading-buffers

Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Order Now

Specificaties

Environmentally Preferable
Uiterlijk Blue
Physical form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR