L-Cystine dihydrochloride 98% (by titrimetric analysis on dry substance)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J63717.30 J63717.A1
J63717.30EA 163 EUR
J63717.30 J63717.A1
L-Cystine dihydrochloride 98% (by titrimetric analysis on dry substance)
L-Cystine dihydrochloride

CAS No.: 30925-07-6 Molecular Formula: C6H12N2O4S2.2HCl Formula Weight: 313.22 Physical Form: Powder Appearance: White to pale yellow Melting Point: 228-232° MDL No.: MFCD00070399

Formule: C₆H₁₂N₂O₄S₂·2HCl
Smeltpunt: 228…232 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00070399
CAS nummer: 30925-07-6
EINECS: 250-391-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR