Nα-(p-Toluenesulphonyl)-DL-lysine chloromethyl ketone hydrochloride 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

J63959.MF J63959.MC
J63959.MFEA 95.5 EUR
J63959.MF J63959.MC
Nα-(p-Toluenesulphonyl)-DL-lysine chloromethyl ketone hydrochloride 98%
Nalpha-(p-Toluenesulphonyl)-DL-lysine chloromethyl ketone hydrochloride

Purity: 98% CAS No.: 4238-41-9 Molecular Formula: C14H21ClN2O3S.HCl Formula Weight: 369.31 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white Solubility: Soluble in water MDL No.: MFCD00012513


Applications: Inhibitor of papain and trypsin

Formule: C₁₄H₂₂Cl₂N₂O₃S
MDL Nummer: MFCD00012513
CAS nummer: 4238-41-9
EINECS: 224-199-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR