Gold 95%, nanoparticles 40 nm OD1 (Abs. 530 nm)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J67409.ACEA 95.9 EUR
J67409.AC J67409.AE
Gold 95%, nanoparticles 40 nm OD1 (Abs. 530 nm)
Gold

Purity: 95% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red to pink MDL No.: MFCD00003436


Applications: For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

Formule: Au
MW: 196.97 g/mol
Kookpunt: 2800
Smeltpunt: 1063
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR