Microtube tube racks, foam
Leverancier: JAECE INDUSTRIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FTR100
JAECFTR100EA 197 EUR
JAECFTR100
Microtube tube racks, foam
Rekken Tube Racks
Economical, charcoal gray, open-cell foam rack have a capacity for 50 tubes. Ideal for storage on benchtops, in incubators and refigerators/freezers. Can be used to protect valuable, fragile tubes during mailing/shipping. Ideal alternative to expensive plastic racks.
  • Racks will float for use in waterbaths
  • Stackable