Exsiccatoren, vetvrij
Leverancier: JENCONS SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
250-068
JENC250-068EA 358 EUR
JENC250-068
Exsiccatoren, vetvrij
Exsiccatoren Exsiccatoren, glas
Borosilicate glass. Elastomer ring is compressed under vacuum making a dry seal. The PTFE valve is situated centrally on the lid, which in conjunction with the dry seal, enables the lid to be lifted on and off vertically.
  • Choice of models with or without Seal-Cote plastic protection, which prevents fragmentation of the glass
  • Lids lift off vertically, giving nearly double the storage volume
Certificaten: BS 3423:1962
Leveringsinformatie: Supplied with sealing ring, gauze, and PTFE stopcock. 9 mm O.D. vacuum connection.