Jenway® pH Electrodes
Leverancier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
99960-41 68006-40 59530-70 68006-37 99960-34 10090-75 99960-39 99960-44 90269-17
JENW99960-41EA 340 EUR
JENW99960-41 JENW68006-40 JENW59530-70 JENW68006-37 JENW99960-34 JENW10090-75 JENW99960-39 JENW99960-44 JENW90269-17
Jenway® pH Electrodes
Elektroden pH-elektroden
A variety of pH electrodes for your Jewnay meters.
  • Choose the right electrode for your application
These electrodes are designed for standard measurements of general solutions. The epoxy body makes it especially suitable for field measurements or use in education.