Eco caps
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-9000-0007 JR-070350-10 JR-9000-0013 JR-9000-0014
JOURJR-9000-0007EA 87.8 EUR
JOURJR-9000-0007 JOURJR-070350-10 JOURJR-9000-0013 JOURJR-9000-0014
Eco caps
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
The Eco Cap consists of a three-hole ETFE insert with a polyethylene collar.
  • Compatible with all standard GL 45 thread bottles
  • Also available as bottle kit
There is no threading for fittings on the inside of the tubing ports, the tubing can be easily inserted.
Leveringsinformatie: The Eco Cap is supplied with one cap, polypropylene male Luer plug. The kit also includes safety-coated glass bottle with a option of 1 or 2 L.