Sample loops for Cheminert® valves, PEEK
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-SLC-85-50 JR-SLC-85-500 JR-SLC-85-5 JR-SLC-85-250 JR-SLC-85-20 JR-SLC-85-1000 JR-SLC-85-100 JR-SLC-85-10 JR-SLC-85-200 JR-SLC-85-300
JOURJR-SLC-85-50EA 89.3 EUR
JOURJR-SLC-85-50 JOURJR-SLC-85-500 JOURJR-SLC-85-5 JOURJR-SLC-85-250 JOURJR-SLC-85-20 JOURJR-SLC-85-1000 JOURJR-SLC-85-100 JOURJR-SLC-85-10 JOURJR-SLC-85-200 JOURJR-SLC-85-300
Sample loops for Cheminert® valves, PEEK
Benodigdheden voor chromatografie Injectieloops voor chromatografiesystemen
These sample loops are made from PEEK tubing and fittings. Mechanically strong, chemically inert polymer ideal for HPLC applications. Softener-free polyethylene caps are used to avoid contamination of the tubing.
  • Fits all Cheminert® C1, C2, C2V, and C3 type valves
  • Tolerance of ±0,025 mm
PEEK sample loops are particularly well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems where metal traces may interact with ionic samples or release transition metals that may deactivateMore Product Information