Dust and liquid goggles, Martcare®
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AGC020-301-300 AGC021-201-300
JSPAAGC020-301-300EA 6 EUR
JSPAAGC020-301-300 JSPAAGC021-201-300
Dust and liquid goggles, Martcare®
Ruimzichtbrillen
A cost-effective, compact, and lightweight goggle.
  • Compact and lightweight
  • Tough polycarbonate lens
  • Impact protection
  • Indirect venting offers dust and liquid protection
  • PVC frame
A compact and lightweight goggle with tough polycarbonate lens and PVC frame, offering high resistance to impact from flying particles.
Certificaten: Conforms to EN 166 1.B.