Junior IV goggle
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AGT020-141-300
JSPAAGT020-141-300EA 8.1 EUR
JSPAAGT020-141-300
Junior IV goggle
Ruimzichtbrillen
Junior goggle with reduced size housing, fully conforming to EN 166 34B.
  • Junior goggle
  • Reduced size housing
  • One-piece polycarbonate lens
  • Elastic headband
The lens and body offer protection against medium energy, medium-speed particles at 120 m/s, large dust particles, and liquid droplets. The optical qualities of the lens comply with the requirements of optical class 1 (highest).
Certificaten: EN 166, EN 170.