Industrial safety helmet, EVOLite®
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AJB170-000-100 AJB170-000-200 AJB170-000-500
JSPAAJB170-000-100EA 46.9 EUR
JSPAAJB170-000-100 JSPAAJB170-000-200 JSPAAJB170-000-500
Industrial safety helmet, EVOLite®
Hoofdbescherming Veiligheidshelmen
EVOLite® is one of the lightest safety helmets for sale in the UK, conforming to EN 397.
  • Lightweight industrial safety helmet
  • Tough ABS shell
  • Wheel ratchet adjustment
  • Chamlon sweatband
The EVOLite® Industrial Safety Helmet is lightweight, extremely comfortable, and fits the widest range of head shapes and sizes. The harness and shell are truly integrated as one to give the best performance by keeping it firmly rooted on the head, even when fully inverted. Gives added security and protection when working at height, in high winds, or when there is aMore Product Information