Comfort Plus™ Industrial safety helmet, EVO®3
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AJF160-000-100
JSPAAJF160-000-100EA 18.2 EUR
JSPAAJF160-000-100
Comfort Plus™ Industrial safety helmet, EVO®3
Hoofdbescherming Veiligheidshelmen
The EVO®3 industrial safety helmet combines a super strong shell for superior all day protection, with the comfort benefits of the Evolution® 3D Adjustment™ harness system
  • Industrial safety helmet
  • Tough HDPE shell
  • 6-point terylene cradle harness
  • Prescision fit
  • Vented design
  • OneTouch™ slip ratchet adjustment
The EVO®3 Comfort Plus™ helmet combines a super strong shell for superior all day protection in the widest range of environments, with the comfort benefits of the Evolution® 3D Adjustment™ harness system. In tests the EVO®3 proved far tougher thanMore Product Information