Adapters, sperical joints
Leverancier: JUCHHEIM Laborgerate GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20457.35 20457.51
JUCH20457.35EA 728 EUR
JUCH20457.35 JUCH20457.51
Adapters, sperical joints
Adapters Adapters met kogelgewrichten
For reaction vessels, 316 stainless steel, thread length 14 mm.