Drying trays
Leverancier: JUCHHEIM Laborgerate GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VLSOBAYB807801 VLSOBAYB807821 VLSOBAYB807841 VLSOBAYB807861
JUCHVLSOBAYB807801EA 122.8 EUR
JUCHVLSOBAYB807801 JUCHVLSOBAYB807821 JUCHVLSOBAYB807841 JUCHVLSOBAYB807861
Drying trays
Reservoirs/Schalen/Potten Droogschalen
Made of acid resistant stainless steel.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
Order Now
Specificaties
Environmentally Preferable