Gel extraction kit, GeneJET

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

K0691 K0692
K0691EA 109 EUR
K0691 K0692
Gel extraction kit, GeneJET
Reagentia voor nucleïnezuren Nucleic Acid Purification Kits and Reagents

GeneJET Gel Extraction Kit is designed for rapid and efficient purification of DNA fragments from standard or low-melting point agarose gels run in either TAE or TBE buffer.


  • Highly efficient up to 95% recoveries in the range of 100 bp to 10 kb
  • Procedure takes only 15 minutes
  • Spin columns are capped and assembled with collection tubes


The kit utilises a proprietary silica-based membrane technology in the form of a convenient spin column. The kit can be used to purify DNA fragments from 25 bp to 20 kb in size with recovery rates up to 95%. Each GeneJET purification column has a binding capacity of up to 25 µg of DNA and can process up to 1 g of agarose gel. The entire procedure takes just 15 minutes, and the isolated DNA is ready to use in all common downstream applications, including ligation, restriction digestion, PCR, sequencing, and labeling.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR