VWR® UltraPure, Tris-acetate-EDTA (TAE) buffer
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
K915-1.6LEA 192 EUR
K915-1.6L K915-4L K915-5L
VWR® UltraPure, Tris-acetate-EDTA (TAE) buffer
Buffers Buffers voor elektroforese Running-buffers
TAE is an extensively used buffer for agarose gel electrophoresis applications requiring high resolution and separation of high molecular weight, double-stranded DNA. TAE buffer is more compatible with in-gel manipulations and band recovery procedures than TBE buffer.
  • Ideal for DNA recovery and in-gel manipulations
  • Low ionic strength
  • Better resolution of large DNA fragments (>12 kb DNA)
  • Convenient packaging options minimise weighing and mixing
Composition of (1X) TAE buffer: 0,04 M Tris-acetate; 0,001 M EDTA; final pH 8,0.
Order Now
Specificaties
Environmentally Preferable