Methanol ≥99.8% for HPLC

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

K977-1LEA 150 EUR
K977-1L K977-4L
Methanol ≥99.8% for HPLC
Methanol
Formule: CH₃OH
Kookpunt: 64,6 °C (1013 hPa)
Smeltpunt: –98 °C
Dichtheid: 0,7918 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
Bereik: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs. at 210 nm <0.8
Abs. at 220 nm ≤0.4
Abs.@230 nm ≤0.2
Abs. at 240 nm ≤0.1
Abs. at 260 nm ≤0.04
Abs.@280 – 400 nm ≤0.01
Carbonyl Compounds ≤0.001%
Color (APHA) ≤10
Purity ≥99.8%
Residue After Evaporation ≤0.001%
Solubility (30%, Water) Pass
Substances Darkened by Sulphuric Acid Pass
Substances Reducing Permanganate Test Pass
Titrable Acid ≤0.0003 meq/g
Titrable Base ≤0.0002 meq/g
Water ≤0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR