DuPont™ Tyvek® 1073B Self-Sealing Autoclave Bags with Steam Indicator
Leverancier: KEYSTONE CLEANROOM PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
COVMGTVK68SS
KACCCOVMGTVK68SSEA 594 EUR
KACCCOVMGTVK68SS
DuPont™ Tyvek® 1073B Self-Sealing Autoclave Bags with Steam Indicator
Zakken Sterilisatiezakken
Self-sealing 1073B Tyvek® are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps as well as heavy equipment parts with sharp edges. No need to use or validate a heat sealing machine.
  • Manufactured and packaged in a controlled cleanroom environment
  • Convenient Self-sealing closure system
  • Steam sterilization indicator - Autoclavable at 121 to 125 °C
  • Tear and puncture resistant, hydrophobic, lint-free
  • 100% Medical Grade 1073B Tyvek® permits fast steam penetration
  • Excellent impact resistance, exceptional