Centrifugeerbuisjes, conische bodem
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
84008 84013 84006 84007 84011 84012 84009 84010 84015 84014
KART84008EA 401 EUR
KART84008 KART84013 KART84006 KART84007 KART84011 KART84012 KART84009 KART84010 KART84015 KART84014
Centrifugeerbuisjes, conische bodem
Buisjes Centrifugebuisjes
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Graduated
Conical bottom with tight screw cap.