Flessen, smalle hals, rond, met schroefdraad
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
302-70515 302-70517 302-70512 302-70513
215-2345EA 3 EUR
215-2345 215-2346 KAUT302-70512 KAUT302-70513
Flessen, smalle hals, rond, met schroefdraad
Flessen Kunststof flessen
LDPE. Round. Natural colour.
  • High narrow shape
  • Easy handling
  • Leakproof
  • Suitable for food
  • Halal certified
Three types of closures for different applications. Ideal for liquids. Handy shape.
Bestelinformatie: The bottles are supplied without closures, these should be ordered separately.