Funnels, powder
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
632413102100 632413102060 632413102080
KAVA632413102100EA 46.1 EUR
KAVA632413102100 KAVA632413102060 KAVA632413102080
Funnels, powder
Trechters Poedertrechters
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These funnels with short stem and bevelled edges are suitable for transferring powdery materials.
Bestelinformatie: Ø = internal stem diameter.