Erlenmeyer flasks with screw cap
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
632426731100 632426731940
KAVA632426731100EA 461 EUR
KAVA632426731100 KAVA632426731940
Erlenmeyer flasks with screw cap
Kolven Erlenmeyers
Borosilicate glass 3,3, clear, with teflon lined blue PP screw cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. Cap temperature resistant up to 140 °C.