Industrial gloves, Camapur® Comfort 616
Leverancier: KCL by Honeywell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
616.8 616.7 616.9 616.10 616.11 616.6
KCHL616.8EA 86.6 EUR
KCHL616.8 KCHL616.7 KCHL616.9 KCHL616.10 KCHL616.11 KCHL616.6
Industrial gloves, Camapur® Comfort 616
Handschoenen
These seamless polyamide liners feature a PU palm coating which provides excellent grip when handling greasy and oily parts.
  • Excellent fit
  • Very breathable
  • Good mechanical resistance
  • Allows working without user fatigue
  • Less sensitive to soiling
  • Washable
Ideal for usage in the electronics and automotive industry, assembly works, for precision mechanics, protection of products, metal and plastics processing and for storage and commission work.
Certificaten: EN 388 (3131); Cat. II