Blood collection tubes with sodium citrate, EMOPLASTIK®
Leverancier: KIMA SAS DI CHIARIN RENZO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
18614
KIMA18614EA 0 EUR
KIMA18614
Blood collection tubes with sodium citrate, EMOPLASTIK®
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PP, translucent.
  • 0,25 ml of sodium citrate
  • Non sterile
  • With writing area