Osmometer, semi-micro, K-7400S
Leverancier: KNAUER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A0006AC
KNAUA0006ACEA 7610 EUR
KNAUA0006AC
Osmometer, semi-micro, K-7400S
Diagnostische systemen Clinical Analysers
The K-7400S semi-mirco osmometer enables the measurement of osmolality as well as the freezing point of aqueous solutions. The proven technology of freezing point determination in combination with the robust and intelligent design of the device allows fast and reproducible measurements.
  • Two-point or optional three-point calibration
  • LED display of two rows with 24 characters
The instrument is equipped with a peltier cooler and an integrated microprocessor controlling the automated measurement. The freezing point osmometer is a standalone device thatMore Product Information