Diaphragm vacuum pumps, LABOPORT®
Leverancier: KNF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N938.50KT.18
KNFAN938.50KT.18EA 1497 EUR
KNFAN938.50KT.18
Diaphragm vacuum pumps, LABOPORT®
Pompen Membraanvacuümpompen
This diaphragm pumps is double-head, dry-running devices used in a wide range of laboratory applications.
Materials: PPS pump head; PTFE-coated diaphragm; FFPM valves.