Transport and Storage Boxes, PP
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2-1145
KOEH2-1145EA 91.8 EUR
KOEH2-1145
Transport and Storage Boxes, PP
Dozen Transportdozen
Transport and storage box made of grey PP. Closed bottom and sides.
  • According to Euronorm measures
  • Resistant against acids and basics
  • Temperature resistant from –30 to 80 °C
  • Stackable