Permanent marker
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2-5064 2-5071 2-5068
KOEH2-5064EA 16.1 EUR
KOEH2-5064 KOEH2-5071 KOEH2-5068
Permanent marker
Pennen Permanente markeerstiften
This versatile paint marker dispense a fast-drying, high-gloss enamel paint for permanent marking, lettering, coding and numbering of both interior and exterior surfaces of metal, plastic, printed circuit boards, glass, wood, paper, leather, textiles, etc. This marker can write on porous or non-porous; rough or smooth; wet, oily or dry surfaces.
  • Marks are permanent
  • Withstand heat and weathering under adverse conditions
  • Available in different lead-free colours
  • With long-lasting replaceable fiber tips
Order Now
Specificaties
Environmentally Preferable