Wall-mounted cabinets
Leverancier: KOETTERMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
307-00021 307-00064 307-00065 307-00066 307-00067 307-00068 307-00069 307-00070 307-00071 307-00072 307-00073 307-00074 307-00075 307-00151 307-00023 307-00024 307-00025 307-00026 307-00027 307-00028 307-00029 307-00030 307-00154 307-00152 307-00020 307-00155 307-00156 307-00153 307-00022 307-00013 307-00014 307-00015 307-00016 307-00017 307-00018 307-00019
KOTT307-00021EA 1020 EUR
KOTT307-00021 KOTT307-00064 KOTT307-00065 KOTT307-00066 KOTT307-00067 KOTT307-00068 KOTT307-00069 KOTT307-00070 KOTT307-00071 KOTT307-00072 KOTT307-00073 KOTT307-00074 KOTT307-00075 KOTT307-00151 KOTT307-00023 KOTT307-00024 KOTT307-00025 KOTT307-00026 KOTT307-00027 KOTT307-00028 KOTT307-00029 KOTT307-00030 KOTT307-00154 KOTT307-00152 KOTT307-00020 KOTT307-00155 KOTT307-00156 KOTT307-00153 KOTT307-00022 KOTT307-00013 KOTT307-00014 KOTT307-00015 KOTT307-00016 KOTT307-00017 KOTT307-00018 KOTT307-00019
Wall-mounted cabinets
Meubilair Opslagkasten
Made of epoxy powder coated steel, with one shelf.
  • Solid hinged doors or sliding glass doors
  • Height adjustable shelfs
  • Maximum loading capacity: 80 kg
Order Now
Specificaties
Environmentally Preferable