Balance benches
Leverancier: KOETTERMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
509-00000 509-00001
KOTT509-00000EA 2520 EUR
KOTT509-00000 KOTT509-00001
Balance benches
Meubilair Tafels
These balance benches are designed to maintain vibration free condition, so that even small quantities can be weighed accurately.
Made from epoxy powder coated steel.