Silver p-toluenesulphonate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L00307.06 L00307.14
L00307.06EA 44.3 EUR
L00307.06 L00307.14
Silver p-toluenesulphonate ≥98%
Silver p-toluenesulphonate

Purity: 98+% CAS No.: 16836-95-6 Molecular Formula: C7H7AgO3S Formula Weight: 279.07 Odour: Odourless UN No.: 3077 MDL No.: MFCD00007511

Formule: C₇H₇AgO₃S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007511
CAS nummer: 16836-95-6
EINECS: 240-859-0
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR